Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecanedioic acid, 99%

Catalog No. p-7080734
Change view
Click to view available options
Quantity:
250 g
1000 g
Catalog No. Quantity
AAA1038730 250 g
AAA103870B 1000 g
2 options

Catalog No. AAA1038730

Supplier: Thermo Scientific Chemicals A1038730

Only null left
Add to Cart

CAS: 693-23-2 | C12H22O4 | 230.304 g/mol

Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.

Solubility
Soluble in water, ethanol, hot methanol, hot toluene and hot acetic acid.

Notes
Incompatible with strong oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-23-2
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00002735
InChI Key TVIDDXQYHWJXFK-UHFFFAOYSA-N
Synonym 1,12-dodecanedioic acid, 1,10-decanedicarboxylic acid, decamethylenedicarboxylic acid, 1,10-dicarboxydecane, n-dodecanedioic acid, unii-978yu42q6i, dsstox_cid_7297, dsstox_rid_78394, dsstox_gsid_27297, n-dodecanedioate
PubChem CID 12736
ChEBI CHEBI:4676
IUPAC Name dodecanedioic acid
SMILES C(CCCCCC(=O)O)CCCCC(=O)O

Specifications

Melting Point 127°C to 131°C
Density 1.15
Boiling Point 244°C to 245°C (10 mmHg)
Flash Point 227°C (440°F)
Odor Odorless
Quantity 250 g
Beilstein 1782580
Solubility Information Soluble in water,ethanol,hot methanol,hot toluene and hot acetic acid.
Formula Weight 230.31
Percent Purity 99%
Chemical Name or Material Dodecanedioic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-746-3
RTECSNumber JR2370000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.