Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyl Gallate, TCI America™
SDP

Catalog No. g001525g
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 1166-52-5
Molecular Formula C19H30O5
Molecular Weight (g/mol) 338.444
MDL Number MFCD00002195
InChI Key RPWFJAMTCNSJKK-UHFFFAOYSA-N
Synonym dodecyl gallate, lauryl gallate, nipagallin la, progallin la, gallic acid, dodecyl ester, gallic acid, lauryl ester, n-dodecyl gallate, n-dodecylgallate, benzoic acid, 3,4,5-trihydroxy-, dodecyl ester, gallic acid dodecyl ester
PubChem CID 14425
IUPAC Name dodecyl 3,4,5-trihydroxybenzoate
SMILES CCCCCCCCCCCCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 25 g
Formula Weight 338.44
Physical Form Crystalline Powder
Chemical Name or Material Dodecyl Gallate
EINECSNumber (3)-1584
RTECSNumber DH9100000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.