Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylamine 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
Packaging:
Glass bottle
124-22-1
C12H27N
185.35
MFCD00008154
JRBPAEWTRLWTQC-UHFFFAOYSA-N
dodecylamine,1-dodecanamine,laurylamine,1-aminododecane,n-dodecylamine,dodecanamine,1-dodecylamine,lauramine,n-laurylamine,alamine 4
13583
dodecan-1-amine
CCCCCCCCCCCCN
124-22-1
C12H27N
185.35
MFCD00008154
JRBPAEWTRLWTQC-UHFFFAOYSA-N
dodecylamine,1-dodecanamine,laurylamine,1-aminododecane,n-dodecylamine,dodecanamine,1-dodecylamine,lauramine,n-laurylamine,alamine 4
13583
dodecan-1-amine
CCCCCCCCCCCCN
Packaging Glass bottle
Color White
Density 0.8
Flash Point >110°C
Quantity 1kg
Infrared Spectrum Authentic
Melting Point 28.0°C to 32.0°C
Boiling Point 247.0°C to 249.0°C
Assay Percent Range 98%
Linear Formula CH3(CH2)11NH2
Beilstein 04, 200
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, isopropylalcohol, ethanol,, ether
Formula Weight 185.35
Physical Form Crystalline Low Melting Solid
Percent Purity 98%
Chemical Name or Material Dodecylamine, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-690-6

RTECSNumber : JR6475000

TSCA : TSCA