Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dodecylamine, 98%
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 28.0°C to 32.0°C |
| Color | White |
| Density | 0.8 |
| Boiling Point | 247.0°C to 249.0°C |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (Alkyl chain distribution C12) (GC) |
| Packaging | Glass bottle |
| Linear Formula | CH3(CH2)11NH2 |
| Quantity | 500 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause respiratory irritation. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquat |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with wat |
| EINECSNumber | 204-690-6 |
| RTECSNumber | JR6475000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction