Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylamine, 98%

Catalog No. p-124473
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle

CAS: 124-22-1 | C12H27N | 185.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-22-1
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.36
MDL Number MFCD00008154
InChI Key JRBPAEWTRLWTQC-UHFFFAOYSA-N
Synonym dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4
PubChem CID 13583
IUPAC Name dodecan-1-amine
SMILES CCCCCCCCCCCCN

Specifications

Melting Point 28.0°C to 32.0°C
Color White
Density 0.8
Boiling Point 247.0°C to 249.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Alkyl chain distribution C12) (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)11NH2
Quantity 100 g
Beilstein 04, 200
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, isopropylalcohol, ethanol,, ether
Formula Weight 185.35
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Dodecylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquat
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 204-690-6
RTECSNumber JR6475000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.