Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylbenzene Sodium Sulfonate, Powder, Approx. 90%, Spectrum™ Chemical
SDP

Catalog No. 18602319 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
12 kg
25 kg
90 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-319 25 kg Fiber Drum
18-602-315 100 g Poly Bottle
18-602-316 500 g Poly Bottle
18-602-317 3 kg Poly Pail
18-602-318 12 kg Poly Pail
18-602-320 90 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-319 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D106425KGBL
Add to Cart
Add to Cart

D1064, 25155-30-0, C18H29NaO3S

Spectrum™ Chemical Dodecylbenzene Sodium Sulfonate, Powder

Chemical Identifiers

CAS 25155-30-0
Molecular Formula C18H29NaO3S
Molecular Weight (g/mol) 348.48
InChI Key HFQQZARZPUDIFP-UHFFFAOYSA-M
IUPAC Name sodium 2-dodecylbenzene-1-sulfonate
SMILES [Na+].CCCCCCCCCCCCC1=C(C=CC=C1)S([O-])(=O)=O

Specifications

CAS Max % 100%
Packaging Fiber Drum
Quantity 25 kg
Percent Purity ∼90%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.