Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyldimethylchlorosilane, 97%

Catalog No. p-4228639
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle

CAS: 66604-31-7 | C14H31ClSi | 262.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66604-31-7
Molecular Formula C14H31ClSi
Molecular Weight (g/mol) 262.93
InChI Key DLLABNOCKQMTEJ-UHFFFAOYSA-N
Synonym dodecyldimethylchlorosilane, chloro dodecyl dimethylsilane, 1-chlorododecyl-dimethyl-silicon, chlorododecyldimethylsilane, acmc-209nvw, dimethyldodecylchlorosilane, dimethyldodecylsilyl chloride, n-dodecyldimethylchlorosilane, dimethyl-dodecyl-chlorosilane, chloro-dimethyl-dodecylsilane
PubChem CID 2758731
IUPAC Name chloro-dodecyl-dimethylsilane
SMILES CCCCCCCCCCCC[Si](C)(C)Cl

Specifications

Color Colorless
Density 0.8650g/mL
Boiling Point 291.0°C to 293.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4440 to 1.4460 (20°C, 589nm)
Linear Formula Cl(CH3)2Si(CH2)11CH3
Quantity 25 mL
Specific Gravity 0.865
Solubility Information Solubility in water: insoluble.
Formula Weight 262.93
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dodecyldimethylchlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 266-421-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.