Learn More
Invitrogen™ DOG-1 Recombinant Rabbit Monoclonal Antibody (SP31)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA516358
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is gastrointestinal stromal tumor.
DOG-1, the novel marker, is expressed ubiquitously in gastrointestinal stromal tumors irrespective of kit or PDGFR alpha mutation status.
Specifications
DOG-1 | |
Recombinant Monoclonal | |
0.055 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.2 | |
Q5XXA6 | |
ANO1 | |
Synthetic peptides of human DOG-1 protein (aa 400-500). | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
SP31 | |
Unconjugated | |
ANO1 | |
ANO1; anoctamin 1; anoctamin 1, calcium activated chloride channel; anoctamin-1; Ca2+-activated Cl- channel; calcium activated chloride channel; discovered on gastrointestinal stromal tumors protein 1; DOG1; FLJ10261; oral cancer overexpressed 2; Oral cancer overexpressed protein 2; ORAOV2; TAOS2; Tmem16a; Transmembrane protein 16A; transmembrane protein 16A (eight membrane-spanning domains); tumor-amplified and overexpressed sequence 2 | |
Rabbit | |
Protein A | |
RUO | |
55107 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.