Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T Lymphocyte Cocktail Mouse anti-Dog, APC, FITC, PE, Clone: LSM 8.358, LSM 11.425, CTL 2.58, BD

Catalog No. BDB558704 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558704 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558704 Supplier BD Biosciences Supplier No. 558704
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cocktail Component Clone Isotype

APC anti-Dog Pan T Cell marker LSM 8.358 mIgM

FITC anti-Dog T cell Activation marker CTL 2.58 mIgG1

The Dog Lymphocyte Activation Cocktail is a three-color reagent cocktail designed to identify dog activated lymphocytes by direct immunofluorescent staining with flow cytometric analysis. The LSM 8.358 antibody reacts with a CD3-like T-cell receptor-associated cell-surface antigen found on thymocytes and peripheral T lymphocytes.The LSM 11.425 antibody was generated against cells derived from canine peripheral lymph nodes and has been used as a prognostic tool in dog B cell lymphoma studies. Clone CTL 2.58 was generated by using whole cell immunizations of IL-2 dependent feline T cell lines stimulated with PHA and Con A. This clone reacts with dog and is referred to as the T Cell Activation Marker.

TRUSTED_SUSTAINABILITY

Specifications

Antigen T Lymphocyte Cocktail
Applications Western Blot
Classification Monoclonal
Clone LSM 8.358, LSM 11.425, CTL 2.58
Conjugate FITC/APC/PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1, IgM
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.