Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOG1 Monoclonal Antibody (ZY210), Abbomax

Catalog No. 502204387
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4387 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4387 Supplier Abbomax Inc Supplier No. 605780
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Human GIST tumor Cellular location: Membrane, Cytoplasm.

DOG1 Monoclonal antibody specifically detects DOG1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen DOG1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ZY210
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene ANO1
Gene Accession No. Q5XXA6
Gene Alias ANO1; anoctamin 1; anoctamin 1, calcium activated chloride channel; anoctamin-1; Ca2+-activated Cl- channel; calcium activated chloride channel; discovered on gastrointestinal stromal tumors protein 1; DOG1; FLJ10261; oral cancer overexpressed 2; Oral cancer overexpressed protein 2; ORAOV2; TAOS2; Tmem16a; Transmembrane protein 16A; transmembrane protein 16A (eight membrane-spanning domains); tumor-amplified and overexpressed sequence 2
Host Species Mouse
Immunogen A partial recombinant protein of human DOG1
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55107
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.