Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOG1/TMEM16A Antibody (006), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289358JF549
Description
DOG1/TMEM16A Monoclonal antibody specifically detects DOG1/TMEM16A in Human samples. It is validated for Western BlotSpecifications
| DOG1/TMEM16A | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 55107 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| 006 | |
| Western Blot | |
| ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A | |
| This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the C-terminus of the Human DOG1/TMEM16A. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction