Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOG1/TMEM16A Antibody (DG1/1484), Novus Biologicals™

Mouse Monoclonal Antibody
$203.51 - $480.74
Specifications
| Antigen | DOG1/TMEM16A |
|---|---|
| Clone | DG1/1484 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25315720
![]() |
Novus Biologicals
NBP25315720UG |
20 μg |
Each for $203.51
|
|
|||||
NB253157100
![]() |
Novus Biologicals
NBP253157100UG |
100 μg |
Each for $480.74
|
|
|||||
NB253157200
![]() |
Novus Biologicals
NBP253157200UG |
200 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
DOG1/TMEM16A Monoclonal specifically detects DOG1/TMEM16A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| DOG1/TMEM16A | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| Unconjugated | |
| Mouse | |
| Human | |
| ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A | |
| TMEM16A | |
| IgG2b κ | |
| Protein A or G purified | |
| 114 kDa |
| DG1/1484 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 55107 | |
| Recombinant human DOG-1 protein fragment (aa 2-101) (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title