Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOG1/TMEM16A Antibody (DG1/1486), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$243.50 - $487.50

Specifications

Antigen DOG1/TMEM16A
Clone DG1/1486
Concentration 0.2 mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25316320
SDP
View Documents
Novus Biologicals
NBP25316320UG
20 μg
Each for $243.50
Only null left
Add to Cart
 
NB253163100
SDP
View Documents
Novus Biologicals
NBP253163100UG
100 μg
Each for $487.50
Only null left
Add to Cart
 
NB253163200
SDP
View Documents
Novus Biologicals
NBP253163200UG
200 μg N/A N/A N/A
Description

Description

DOG1/TMEM16A Monoclonal specifically detects DOG1/TMEM16A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

DOG1/TMEM16A
0.2 mg/mL
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
Unconjugated
Mouse
Human
ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A
TMEM16A
IgG2b κ
Protein A or G purified
114 kDa
DG1/1486
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page
Monoclonal
Purified
RUO
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
55107
Recombinant human DOG-1 protein fragment (aa 2-101) (exact sequence is proprietary)
Primary
Store at 4C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.