Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOG1/TMEM16A Antibody (DG1/2564R), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308701AF532
Description
DOG1/TMEM16A Monoclonal antibody specifically detects DOG1/TMEM16A in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin)Specifications
DOG1/TMEM16A | |
Monoclonal | |
Alexa Fluor 532 | |
ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A | |
A recombinant human canine-1 protein fragment (around aa 2-101) (exact sequence is proprietary) (Uniprot: Q5XX6) | |
100 μg | |
Primary | |
Human, Canine | |
Purified |
Immunohistochemistry (Paraffin) | |
DG1/2564R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
55107 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction