Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOG1/TMEM16A Antibody (OTI1C8), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271987AF700
Description
DOG1/TMEM16A Monoclonal specifically detects DOG1/TMEM16A in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
DOG1/TMEM16A | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ANO1 | |
Recombinant protein fragment corresponding to amino acids 1-333 of human DOG1/TMEM16A (NP_060513) produced in E.coli. | |
0.1 mL | |
55107.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI1C8 | |
Immunohistochemistry | |
ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction