Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOG1/TMEM16A Antibody (OTI1C8), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271987PE
Description
DOG1/TMEM16A Monoclonal antibody specifically detects DOG1/TMEM16A in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DOG1/TMEM16A | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
55107 | |
Store at 4C in the dark. | |
IgG1 |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI1C8 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A | |
Human recombinant protein fragment corresponding to amino acids 1-333 of human ANO1 (NP_060513) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction