Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOK1, p Ser450 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323286
Description
DOK1 Polyclonal antibody specifically detects DOK1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
DOK1 | |
Polyclonal | |
Western Blot 1:1000 | |
docking protein 1, docking protein 1 (downstream of tyrosine kinase 1), docking protein 1, 62kD (downstream of tyrosine kinase 1), docking protein 1, 62kDa (downstream of tyrosine kinase 1), Downstream of tyrosine kinase 1, MGC117395, MGC138860, p62(dok), P62DOK, pp62 | |
Phospho-Dok1 (Ser-450) synthetic peptide (coupled to KLH) corresponds to amino acids surrounding serine 450 in human Dok1. This sequence is conserved in Dok1 from rat (Ser-449) and mouse (Ser-451). The site is not conserved in other Dok family members. [UniProt# Q99704] | |
0.1 mL | |
Phospho Specific, Signal Transduction | |
1796 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction