Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOK1, p Tyr362 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$243.50 - $402.00

Specifications

Antigen DOK1 (p Tyr362)
Concentration 1.0 mg/mL
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017870
SDP
View Documents
Novus Biologicals
NB100821120.025ML
0.025 mL
Each for $243.50
Only null left
Add to Cart
 
NB10082112
SDP
View Documents
Novus Biologicals
NB10082112
0.05 mL
Each for $402.00
Only null left
Add to Cart
 
Description

Description

DOK1 Polyclonal specifically detects DOK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DOK1 (p Tyr362)
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Polyclonal
Rabbit
Phospho Specific, Signal Transduction
Q99704
1796
IgG
Affinity Purified
62 kDa
1.0 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Unconjugated
RUO
Mouse, Rat, Human
docking protein 1, docking protein 1 (downstream of tyrosine kinase 1), docking protein 1, 62kD (downstream of tyrosine kinase 1), docking protein 1, 62kDa (downstream of tyrosine kinase 1), Downstream of tyrosine kinase 1, MGC117395, MGC138860, p62(dok), P62DOK, pp62
DOK1
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.