Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOK1 Recombinant Rabbit Monoclonal Antibody (JE54-52)
GREENER_CHOICE

Catalog No. PIMA536191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536191 Supplier Invitrogen™ Supplier No. MA536191
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

DOK1 Recombinant Monoclonal Antibody for Western Blot

DOK1 (P62dok, Docking protein 1) is believed to be a mainly cytoplasmic adaptor protein which down-regulates mitogen-activated protein kinase activation, inhibits cell proliferation and transformation, and promotes cell spreading and cell migration. DOK1 is a major substrate for many tyrosine kinases including c-kit, v-abl, vFPS, EGF, and PDGF. Upon phosphorylation by kinases, DOK1 forms a complex with ras GTPase-activating protein. Phosphorylation on tyrosine residues by the insulin receptor kinase results in the negative regulation of the insulin signaling pathway. DOK1 contains a putative pleckstrin homology domain at the amino terminus and ten PXXP SH3 recognition motifs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOK1
Applications Western Blot
Classification Recombinant Monoclonal
Clone JE54-52
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene DOK1
Gene Accession No. Q4QQV2, Q99704
Gene Alias AW557123; Docking protein 1; docking protein 1, 62kDa (downstream of tyrosine kinase 1); Dok; DOK 1; DOK1; Dok-1; Downstream of tyrosine kinase 1; downstream of tyrosine kinase-1; MGC117395; MGC138860; p240; p62 dok; p62(dok); p62DOK; pp62; TLP1; TP1; TROVE domain family; TROVE1; VAULT2
Gene Symbols DOK1
Host Species Rabbit
Immunogen Recombinant protein within Human DOK1 aa 360-481.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1796, 312477
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.