Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOK2 Antibody (OTI1A4), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271407FR
Description
DOK2 Monoclonal specifically detects DOK2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
DOK2 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
DOK2 | |
Full length recombinant protein of human DOK2 (NP_003965) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1A4 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
docking protein 2, docking protein 2, 56kD, docking protein 2, 56kDa, Dok-2, Downstream of tyrosine kinase 2, p56(dok-2), P56DOK, p56dok-2 | |
Mouse | |
Affinity Purified | |
Phospho Specific, Signal Transduction | |
9046.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction