Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOK2 Monoclonal Antibody (OTI2A3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503302

Description
Docking proteins interact with receptor tyrosine kinases and mediate particular biological responses using signal transduction. Dok-2 acts as a multiple docking protein downstream of receptor or non-receptor tyrosine kinases. By this mechanism it acts to negatively regulate signal transduction and cell proliferation controlled by cytokines in a feedback loop. Dok-2 is highly expressed in cells and tissues of hematopoietic origin as well as in lung. Expression of bcr/abl induces additional tyrosine phosphorylation of the Dok1 and Dok2 proteins and their association with Ras-GAP. Thus, it is suspected that DOK association regulates GAP activity toward Ras and that the Dok proteins serve as mediators of bcr-abl signaling. The role of Dok proteins in bcr-abl regulation may also be implicated in chronic myelogenous leukemia (CML), which is characterized by a Philadelphia chromosome translocation t(9;22). Such a mutation would result in a p210-bcr/abl chimeric protein-tyrosine kinase which has been found in many CML cases.Specifications
DOK2 | |
Monoclonal | |
Unconjugated | |
DOK2 | |
p56DOK, p56dok-2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9046 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2A3 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O60496 | |
DOK2 | |
Full length human recombit protein of human DOK2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction