Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589616 Supplier Invitrogen™ Supplier No. PA589616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGDGAVKQGF LYLQQQQTFG KKWRRFGASL YGGSDCALAR LELQEGPEKP RRCEAARKVI RLSDCLRVAE AGGEASSPRD TSAFFLETKE RLYLLAAPAA ERGDWVQAIC LLAFPGQRKE LSGPEGKQSR PCMEENELYS SAVTVGPHKE FAVTMRPTEA SERCHLRGSY TLRAGESALE LWGGPEPGTQ LYDWPYRFLR RFGRDKVTFS FEAGRRCVSG EGNFEFETRQ GNEIFLALEE AISAQKNAAP ATPQPQPATI; Positive Samples: THP-1, Mouse lung.

The protein encoded by this gene is constitutively tyrosine phosphorylated in hematopoietic progenitors isolated from chronic myelogenous leukemia patients in the chronic phase. It may be a critical substrate for p210(bcr/abl), a chimeric protein whose presence is associated with CML. This encoded protein binds p120 from CML cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOK2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DOK2
Gene Accession No. O60496, O70469
Gene Alias docking protein 2; docking protein 2, 56kD; docking protein 2, 56kDa; Dok2; Dok-2; DokR; Dok-R; dok-related protein; Downstream of tyrosine kinase 2; Frip; IL-Four Receptor Interacting Protein; IL-four receptor-interacting protein; LOW QUALITY PROTEIN: docking protein 2; docking protein 2; p56 dok-2; p56(dok-2); p56DOK; p56dok-2
Gene Symbols DOK2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human DOK2 (NP_0039652).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13449, 9046
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.