Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 157491AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-0451


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The MW of this protein is 53 kDa, and this antibody specifically recognizes the 53 kDa protein.

DOK3 DOK proteins are enzymatically inert adaptor or scaffolding proteins. They provide a docking platform for the assembly of multimolecular signaling complexes. DOK3 is a negative regulator of JNK signaling in B-cells through interaction with INPP5D/SHIP1. May modulate ABL1 function (By similarity).
Specifications

Specifications

DOK3
Polyclonal
Unconjugated
DOK3
docking protein 3, DOK3, DOKL
Rabbit
Antigen Affinity Chromatography
RUO
27261, 306760, 79930
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B2RYG7, Q7L591, Q9QZK7
Dok3
DOK3 Fusion Protein Ag8221
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.