Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOK3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987142
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7142 Supplier Bioss Supplier No. BS6234R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DOK3 DOK proteins are enzymatically inert adaptor or scaffolding proteins. They provide a docking platform for the assembly of multimolecular signaling complexes. DOK3 is a negative regulator of JNK signaling in B-cells through interaction with INPP5D/SHIP1. May modulate ABL1 function (By similarity).

Specifications

Antigen DOK3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DOK3
Gene Accession No. Q7L591
Gene Alias AI450713; Docking protein 3; Dok3; Dokl; DOK-L; Dok-like protein; Downstream of tyrosine kinase 3; FLJ22570; FLJ39939; p62(dok)-like protein; p62Dok-like protein
Gene Symbols DOK3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DOK3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79930
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.