Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563190 Supplier Invitrogen™ Supplier No. PA563190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRLEKYPDEK SVCLRGCPKV TEISNVKCVT RLPKETKRQA VAIIFTDDSA RTFTCDSEL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

DOK proteins are enzymatically inert adaptor or scaffolding proteins. They provide a docking platform for the assembly of multimolecular signaling complexes. DOK4 functions in RET-mediated neurite outgrowth and plays a positive role in activation of the MAP kinase pathway. Putative link with downstream effectors of RET in neuronal differentiation. May be involved in the regulation of the immune response induced by T-cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOK4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DOK4
Gene Accession No. Q8TEW6
Gene Alias docking protein 4; Dok4; Downstream of tyrosine kinase 4; insulin receptor substrate 5; IRS5; IRS-5
Gene Symbols DOK4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DOK4. Recombinant protein control fragment (Product #RP-101658).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55715
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.