Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587381 Supplier Invitrogen™ Supplier No. PA587381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATNFSDIVK QGYVKMKSRK LGIYRRCWLV FRKSSSKGPQ RLEKYPDEKS VCLRGCPKVT EISNVKCVTR LPKETKRQAV AIIFTDDSAR TFTCDSELEA EEWYKTLSVE CLGSRLNDIS LGEPDLLAPG VQCEQTDRFN VFLLPCPNLD VYGECKLQIT HENIYLWDIH NPRVKLVSWP LCSLRRYGRD ATRFTFEAGR MCDAGEGLYT FQTQEGEQIY QRVHSATLAI AEQHKRVLLE MEKNVRLLNK GTEHYSYPCT PTTMLPRSAY WHHITGSQNI AEASSYAGEG YGAAQASSET DLLNRFILLK PKPSQGDSSE AKTPSQ Positive Samples: MCF7.

DOK proteins are enzymatically inert adaptor or scaffolding proteins. They provide a docking platform for the assembly of multimolecular signaling complexes. DOK4 functions in RET-mediated neurite outgrowth and plays a positive role in activation of the MAP kinase pathway. Putative link with downstream effectors of RET in neuronal differentiation. May be involved in the regulation of the immune response induced by T-cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOK4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene DOK4
Gene Accession No. Q8TEW6, Q99KE3
Gene Alias docking protein 4; Dok4; Downstream of tyrosine kinase 4; insulin receptor substrate 5; IRS5; IRS-5
Gene Symbols DOK4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-326 of human DOK4 (NP_0605802).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114255, 361364, 55715
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.