Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOK5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565874
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565874 Supplier Invitrogen™ Supplier No. PA565874
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YWQHITRQHST GQLYRLQDVS SPLKLHRTET FPAYRSEH Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

The protein encoded by this gene is a member of the DOK family of membrane proteins, which are adapter proteins involved in signal transduction. The encoded protein interacts with phosphorylated receptor tyrosine kinases to mediate neurite outgrowth and activation of the MAP kinase pathway. In contrast to other DOK family proteins, this protein does not interact with RASGAP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOK5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DOK5
Gene Accession No. Q9P104
Gene Alias 2700055C10Rik; C20orf180; Docking protein 5; Dok5; Downstream of tyrosine kinase 5; insulin receptor substrate 6; IRS6; IRS-6; MGC16926; Protein dok-5; RGD1562846
Gene Symbols DOK5
Host Species Rabbit
Immunogen Recombinant Human DOK5. Recombinant protein control fragment (Product #RP-106545).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55816
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.