Learn More
Invitrogen™ DOK5 Polyclonal Antibody

Description
Immunogen sequence: LLQSVKNSML QMKMSERAAS LSTMVPLPRS AYWQHITRQH STGQLYRLQD VSSPLKLHRT ETFPAYRSEH; Positive Samples: Mouse kidney, Mouse liver, Rat skeletal muscle.
Specifications
Specifications
| Antigen | DOK5 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.17 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | DOK5 |
| Gene Accession No. | Q91ZM9 |
| Gene Alias | 2700055C10Rik; C20orf180; Docking protein 5; Dok5; Downstream of tyrosine kinase 5; insulin receptor substrate 6; IRS6; IRS-6; MGC16926; Protein dok-5; RGD1562846 |
| Gene Symbols | DOK5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.