Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOK6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559426 Supplier Invitrogen™ Supplier No. PA559426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WHHITRQNSV GEIYSLQGHG FGSSKMSRAQ TFPSYAPEQS EEAQQPLSRS SSYGFSYSSS LIQ Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

DOK-6 (docking protein 6), is a novel member of the Dok-4/5 subclass of the p62 Dok family of intracellular adaptor molecules. Dok-6 is highly expressed in the developing central nervous system and is co-expressed with Ret in several locations, including sympathetic, sensory, and parasympathetic ganglia, as well as in the ureteric buds of the developing kidneys. Pull-down assays using the Dok-6 phosphotyrosine binding (PTB) domain and GDNF-activated Ret indicate that Dok-6 binds to the phosphorylated Ret Tyr(1062) residue. Moreover, ligand activation of Ret resulted in phosphorylation of tyrosine residue(s) located within the unique C terminus of Dok-6 predominantly through a Src-dependent mechanism, indicating that Dok-6 is a substrate of the Ret-Src signaling pathway. (Crowder RJ, Enomoto H, Yang M, Johnson EM Jr, Milbrandt J. Dok-6, a Novel p62 Dok family member, promotes Ret-mediated neurite outgrowth. J Biol Chem. 2004 Oct 1;279(40):42072-81. Epub 2004 Jul 30. It is also known as DOK6, HsT3226, MGC20785 or IRS. 0.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOK6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dok6
Gene Accession No. Q6PKX4
Gene Alias Docking protein 6; DOK5L; Dok6; Dok-6; Downstream of tyrosine kinase 6; downstream of tyrosine kinases 6; HsT3226; RGD1564376
Gene Symbols Dok6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DOK6. Recombinant protein control fragment (Product #RP-98284).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220164
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.