Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOK7 Antibody (OTI1A9), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272494PCP
Description
DOK7 Monoclonal specifically detects DOK7 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| DOK7 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| DOK7 | |
| Full length recombinant protein of human DOK7 (NP_775931) produced in HEK293T cells. | |
| 0.1 mL | |
| 285489.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI1A9 | |
| Flow Cytometry | |
| C4orf25chromosome 4 open reading frame 25, CMS1B, docking protein 7, Dok-7, Downstream of tyrosine kinase 7, FLJ33718, FLJ39137, FLJ90556, protein Dok-7 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction