Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOK7 Antibody (OTI2C12), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272491G
Description
DOK7 Monoclonal specifically detects DOK7 in Human samples. It is validated for Western Blot.Specifications
DOK7 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DOK7 | |
Full length recombinant protein of human DOK7 (NP_775931) produced in HEK293T cells. | |
0.1 mL | |
285489.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2C12 | |
Western Blot | |
C4orf25chromosome 4 open reading frame 25, CMS1B, docking protein 7, Dok-7, Downstream of tyrosine kinase 7, FLJ33718, FLJ39137, FLJ90556, protein Dok-7 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction