Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOK7 Monoclonal Antibody (OTI5C6), TrueMAB™, OriGene
SDP

Catalog No. 502540115
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0115 30 μL
50-167-5403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0115 Supplier OriGene Supplier No. TA504788S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DOK7 Monoclonal antibody specifically detects DOK7 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen DOK7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5C6
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DOK7
Gene Accession No. Q18PE1
Gene Alias A930013K19Rik; AW049091; C4orf25; CMS10; CMS1B; docking protein 7; DOK7; Dok-7; Downstream of tyrosine kinase 7; EF-12; Oit5; oncoprotein induced transcript 5; protein Dok-7; RGD1566416
Host Species Mouse
Immunogen Full length human recombinant protein of human DOK7 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285489
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.