Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOK7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563726
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563726 Supplier Invitrogen™ Supplier No. PA563726
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSLDVWRATD ELGSLLSLPA AGAPEPSLCT CLPGTVEYQV PTSLRAHYDT PRSLCLAPRD HS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.

The protein encoded by this gene is essential for neuromuscular synaptogenesis. The protein functions in aneural activation of muscle-specific receptor kinase, which is required for postsynaptic differentiation, and in the subsequent clustering of the acetylcholine receptor in myotubes. This protein can also induce autophosphorylation of muscle-specific receptor kinase. Mutations in this gene are a cause of familial limb-girdle myasthenia autosomal recessive, which is also known as congenital myasthenic syndrome type 1B. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOK7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DOK7
Gene Accession No. Q18PE1
Gene Alias A930013K19Rik; AW049091; C4orf25; CMS10; CMS1B; docking protein 7; DOK7; Dok-7; Downstream of tyrosine kinase 7; EF-12; Oit5; oncoprotein induced transcript 5; protein Dok-7; RGD1566416
Gene Symbols DOK7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DOK7. Recombinant protein control fragment (Product #RP-101684).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285489
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.