Learn More
Invitrogen™ DOLK Polyclonal Antibody

Description
DOLK Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | DOLK |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Dolk |
| Gene Accession No. | 0 |
| Gene Alias | CDG1M; DK; DK1; dolichol kinase; dolichol kinase 1; DOLK; KIAA1094; SEC59; SEC59 homolog; TMEM15; Transmembrane protein 15; UNQ2422/PRO4980 |
| Gene Symbols | Dolk |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.