Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DOLK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584735
Description
Immunogen sequence: AFYVQYKWDR LLQQGSAVFQ FRMSAN.
Involved in the synthesis of the sugar donor Dol-P-Man which is required in the synthesis of N-linked and O-linked oligosaccharides and for that of GPI anchors.
Specifications
DOLK | |
Polyclonal | |
Unconjugated | |
Dolk | |
CDG1M; DK; DK1; dolichol kinase; dolichol kinase 1; DOLK; KIAA1094; SEC59; SEC59 homolog; TMEM15; Transmembrane protein 15; UNQ2422/PRO4980 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22845 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UPQ8 | |
Dolk | |
Recombinant protein corresponding to Human DOLK. Recombinant protein control fragment (Product #RP-108518). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction