Learn More
Eisco™ Dolostone Sedimentary Rock Specimen
SCI_ED
Catalog No. S27608 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27608 12/Pk.
S27586 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27608 Supplier Eisco™ Supplier No. ESNG0079PK12
Add to Cart
Add to Cart
Dolostone is a lithified form of dolomite. Composed primarily of carbonate material, dolostone is a more stalwart rock than its chemical cousin, limestone. It is often mined for construction and infrastructure purposes.
  • Correlates well to Next Generation Science Standards Evidence
  • Useful for teaching students about sedimentary petrology, chemical sedimentary rocks, and industrial geology
Specifications
Product Type Sedimentary Rock
Grade K to 16
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.07 kg