Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOLPP1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen DOLPP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1599820
SDP
View Documents
Novus Biologicals
NBP15998120UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP159981
SDP
View Documents
Novus Biologicals
NBP159981
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

DOLPP1 Polyclonal specifically detects DOLPP1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

DOLPP1
Polyclonal
Rabbit
Protein Phosphatase
dolichyl pyrophosphate phosphatase 1LSFR2, dolichyldiphosphatase 1, EC 3.6.1, EC 3.6.1.43, linked to Surfeit genes in Fugu rubripes 2
DOLPP1
IgG
Western Blot
Unconjugated
RUO
Q86YN1
57171
Synthetic peptides corresponding to DOLPP1(dolichyl pyrophosphate phosphatase 1) The peptide sequence was selected from the N terminal of DOLPP1. Peptide sequence AADGQCSLPASWRPVTLTHVEYPAGDLSGHLLAYLSLSPVFVIVGFVTLI.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.