Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DOM3Z Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569814
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. The function of its protein product is unknown, but its ubiquitous expression and conservation in both simple and complex eukaryotes suggests that this may be a housekeeping gene.
Specifications
DOM3Z | |
Polyclonal | |
Unconjugated | |
DXO | |
decapping and exoribonuclease protein; decapping exoribonuclease; dom-3 homolog Z; DOM3L; DOM3Z; Dxo; NG6; protein Dom3Z; RAI1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
361799 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6MG77 | |
DXO | |
synthetic peptide directed towards the middle region of human DOM3Z (aa 85-134). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction