Learn More
MCR Safety Dominator™ 3
Shop All MCR Safety Products
                    Click to view available options
                                Lens Tint:
                                Amber
                                Blue
                                Clear
                                Gray
                                Indoor/Outdoor
                                Mirror
                                Frame Color:
                                Black
                                Blue, Yellow
                                Clear
                                Gun Metal
                                Lens Coating:
                                Anti-fog
                                Scratch-resistant
                    Description
- Italian fashion, sport focused design
- Co-injected wire core independently adjustable nose pads
- Co-injected soft wire core adjustable TPR temples
- MAX3™, MAX6™, or BOSSMAN™ Mirror coatings
- Meets or Exceeds Z87+ & Military High Velocity
- Gun Metal/Red TPR, Clear/Blue TPR, Black/Black TPR, or Yellow/Blue TPR
Certifications
CSA
Compliance
Meets ANSI Z87.1-2015, CSA Z94.3 and MIL - PRF - 31013 standards.
Specifications
Specifications
| Lens Tint | Indoor/Outdoor | 
| For Use With (Application) | Manufacturing | 
| Frame Color | Clear | 
| No. per Pack | 1 Pair | 
| Lens Material | Polycarbonate | 
| Frame Material | Polycarbonate | 