Learn More
MilliporeSigma™ Donkey anti-Rabbit IgG, Species Adsorbed: Hu, Ms, Rt, B, G, O, Ck, Gp, H , Secondary Antibody ™ (Chemicon™)
Supplier: MilliporeSigma™ AP182C/DEL
Description
ELISA, Western Blotting
Specifications
Donkey anti-Rabbit IgG, Species Adsorbed: Hu, Ms, Rt, B, G, O, Ck, Gp, H | |
Cyanine3 | |
Affinity Purified | |
RUO | |
Secondary |
Polyclonal | |
Donkey | |
500 μg | |
Enzyme | |
Rabbit |