Learn More
MilliporeSigma™ Donkey anti-Rabbit IgG, Species Adsorbed: Hu, Ms, Rt, B, G, O, Ck, Gp, H , Secondary Antibody ™ (Chemicon™)
Specifications
Antigen | Donkey anti-Rabbit IgG, Species Adsorbed: Hu, Ms, Rt, B, G, O, Ck, Gp, H |
---|---|
Classification | Polyclonal |
Host Species | Donkey |
Regulatory Status | RUO |
Research Discipline | Enzyme |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
50-174-461
|
MilliporeSigma™
AP182C/DEL |
Cyanine3 | 500 μg | N/A | N/A | N/A | |||
50-174-462
|
MilliporeSigma™
AP182F/DEL |
FITC | 500 μg | N/A | N/A | N/A | |||
50-174-464
|
MilliporeSigma™
AP182R/DEL |
Rhodamine | 500 μg | N/A | N/A | N/A | |||
Description
ELISA, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Donkey anti-Rabbit IgG, Species Adsorbed: Hu, Ms, Rt, B, G, O, Ck, Gp, H | |
Donkey | |
Enzyme | |
Secondary |
Polyclonal | |
RUO | |
Rabbit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.