Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey IgG Isotype Control, Rhodamine, Novus Biologicals™
SDP

Catalog No. NBP196909 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP196909 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196909 Supplier Novus Biologicals Supplier No. NBP196909
Add to Cart
Add to Cart

Donkey Antibody

IgG specifically detects IgG in Donkey samples. It is validated for Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.

Specifications

Antigen IgG
Conjugate Rhodamine
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Donkey
Purification Method Multi-step
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Gene ID (Entrez) 3500
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Donkey IgG and anti-Donkey Serum.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Donkey
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.