Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Serum, Donor Horse

Catalog No. mp092921149 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mL

Catalog No. MP092921149

Supplier: MP Biomedicals™ 092921149

Only null left
Add to Cart
Add to Cart

Processed from whole blood collected from a controlled herd of young horses.

  • Donor Horse Serum is suitable for use in diagnostic assays, as a supplement in mycoplasma growth and assay media and for culturing hematopoietic stem cells and neuronal cells.
  • This serum is processed from whole blood collected from a controlled herd of young horses.
  • Each animal is routinely screened for Equine Infectious Anemia (EIA).
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
Form Solution
Product Type Horse Serum
Quantity 100 mL
Concentration Protein: 5.6 to 9.1 g/dL
Sterility Sterile
Sterilization Method 0.2 μm filtration
For Use With (Application) Diagnostic assays
pH 7.1 to 8

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.