Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DONSON Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561865 Supplier Invitrogen™ Supplier No. PA561865
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KARSVNVKTQ ALSGYRDQFS LEITGPIMPH SLHSLTMLLK SSQSGSFSAV LYPHEPTAVF NICLQMDKVL DMEVVHKELT NCGLHPNTLE QLSQIPLLGK SSLRN Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 29%.

DONSON is essential for DNA amplification in the ovary and required for cell proliferation during development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DONSON
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DONSON
Gene Accession No. Q9NYP3
Gene Alias B17; C21orf60; C2TA; DONSON; downstream neighbor of SON; protein downstream neighbor of Son
Gene Symbols DONSON
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DONSON. Recombinant protein control fragment (Product #a href=.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29980
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.