Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dooku 1
GREENER_CHOICESDP

Catalog No. 656850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-685-0 50 mg
65-681-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 65-685-0 Supplier Tocris Bioscience™ Supplier No. 6568/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reversibly blocks Yoda1 activity; does not affect constitutive Piezo1 activity

Dooku 1 reversibly blocks Yoda1 (Cat. No. 5586) activity (IC50 = ∼1.5 μM). Exhibits no effect on constitutive Piezo1 channel activity, but antagonizes Yoda1-induced Piezo1 activation in endothelial cells, and inhibits Yoda1-induced Ca2+-entry in vitro. Also blocks Yoda1-induced relaxation of aorta ex vivo.

Specifications

Chemical Name or Material 2-[(2,6-Dichlorobenzyl)thio)-5-(1 H-pyrrol-2-yl)-1,3,4-oxadiazole
CAS 2253744-54-4
Quantity 50 mg
Target Other Piezo Channel Compounds
Molecular Formula C13H9Cl2N3OS
Formula Weight 326.2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.