Learn More
BINDER™ Door Gasket FKM
Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
Model VD/VDL 115
Model VD/VDL 23
Description
- Option for BINDER series VD/VDL
- Door gasket made of FKM (a fluoroelastomer)
- Silicone free
- Resistant to high temperatures and many chemical substances and is used in cases were silicone fails to meet the specifications.
Specifications
Specifications
Chamber Material | FKM |
For Use With (Equipment) | Model VD/VDL 23 |
Material | FKM |
Type | Door Gasket |