Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Door Lock
SDP

Catalog No. 01672883 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
Units M 56, M 115, M 260
Units M 720

Catalog No. 01-672-883

Supplier: BINDER™ 80122335

Only null left
Add to Cart
Add to Cart

Lockable door handle.

  • Lockable door handle
  • For use with various BINDER™ Unit M lines

Specifications

Description Door Lock
For Use With (Equipment) Units M 56, M 115, M 260
Material Plastic
Weight (English) 2.20 lb.
Weight (Metric) 1 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.