Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Dop1R1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144383
Description
Dop1R1 Polyclonal Antibody for Western Blot, ELISA
Receptor for dopamine. The activity of this receptor is mediated by G proteins which activate adenylyl cyclase. Might be involved in the processing of visual information and/or visual learning. Important for Pavlovian conditioning: required in the mushroom body as a receptor conveying unconditional stimuli information, has a role in memory formation for aversive and appetitive learning. Sleep-deprivation-induced impairments in learning can be partially explained through alterations in dopamine signaling, Dop1R1 expression levels are reduced; sleep may have a role in restoring dopamine homeostasis.
Specifications
| Dop1R1 | |
| Polyclonal | |
| Unconjugated | |
| Dop1R1 | |
| DopR; DopR1; DopR35EF | |
| Rabbit | |
| Protein G | |
| RUO | |
| 41726 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4.486 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P41596 | |
| Dop1R1 | |
| Recombinant Drosophila melanogaster (Fruit fly) Dop1R1 protein (20-142aa ). | |
| 100 μL | |
| Primary | |
| Drosophila | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction