Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dopa Decarboxylase/DDC Antibody (40), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289567MFV500
Description
Dopa Decarboxylase/DDC Monoclonal antibody specifically detects Dopa Decarboxylase/DDC in Human samples. It is validated for ELISA, ELISASpecifications
Dopa Decarboxylase/DDC | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Sandwich ELISA | |
40 | |
ELISA, Sandwich ELISA Detection | |
AADCaromatic-L-amino-acid decarboxylase, DOPA decarboxylase, dopa decarboxylase (aromatic L-amino acid decarboxylase), EC 4.1.1, EC 4.1.1.28 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Dopa Decarboxylase/DDC (Accession#: NP_000781.1; Met 1-Glu 480). | |
0.1 mL | |
Neuroscience | |
1644 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction