Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Dopa Decarboxylase/DDC Antibody Pair [HRP]
SDP

Catalog No. NB010030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010030 5 Plates
NB010029 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB010030 Supplier Novus Biologicals™ Supplier No. NBP2794955PLATES
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Dopa Decarboxylase/DDC.

Specifications

Assay Range 109.38 pg/mL
Assay Sensitivity 156.25-10000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Dopa Decarboxylase/DDC.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias AADCaromatic-L-amino-acid decarboxylase, DOPA decarboxylase, dopa decarboxylase (aromatic L-amino acid decarboxylase), EC 4.1.1, EC 4.1.1.28
Gene ID (Entrez) 1644
Gene Symbol DDC
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human Dopa Decarboxylase/DDC monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 2 mg/mL of rabbit anti-Human Dopa Decarboxylase/DDC monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 400 ng of recombinant Human Dopa Decarboxylase/DDC.
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Neuroscience
    Storage Requirements Storage is content dependent.
    Target Dopa Decarboxylase/DDC
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.2 μg/mL in Dilution Buffer before use. 3. Standard: Reconstitute with 1 ml Dilution Buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 10000 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.